Learn More
American Research Products Inc Rabbit Pulmonary surfactant as
Catalog No. 5000327741
Encompass
Rabbit Pulmonary surfactant-associated protein A (SFTPA1) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27741 Supplier American Research Products Inc Supplier No. CSBEL021168RB
Rabbit Pulmonary surfactant-associated protein A (SFTPA1) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich