Learn More
American Research Products Inc Rabbit Pulmonary surfactant as
Supplier:  American Research Products Inc CSBEL021173RB
Encompass
Rabbit Pulmonary surfactant-associated protein B (SFTPB) ELISA kit from CUSABIO Detection Range 1 87 ng/mL-120 ng/mL Sensitivity 0 46 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27742