Learn More
Biomatik Corporation Rabbit S100 Calcium Binding Protein A11 (S100A11) ELISA Kit
Supplier:  Biomatik Corporation EKU11410
Encompass_Preferred
Rabbit S100 Calcium Binding Protein A11 (S100A11) ELISA Kit for detecting S100A11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P24480. Gene ID: 100008975. Target Synonyms: S100-A11; S100C; MLN70; Protein S100-C; Calgizzarin; Metastatic lymph node gene 70 protein; Protein S100-A11, N-terminally processed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5824