Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Rabbit Serum

Catalog No. ICN19135780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 mL
100 mL
Form:
Liquid
Concentration:
4 to 7 g/dL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form Concentration
ICN19135780 100 mL Liquid 4 to 7 g/dL
ICN19135720 20 mL Liquid 4 to 7 g/dL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19135780 Supplier MP Biomedicals™ Supplier No. 0219135780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sterile, Frozen Brown Solution. Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance U.S. regulations
Content And Storage -20°C
Species Rabbit
Form Liquid
Product Type Rabbit Serum
Quantity 100 mL
Concentration 4 to 7 g/dL
Sterility Sterile
Sterilization Method Membrane-filtered
For Use With (Application) Blocking agent in immunohistochemistry culture media component
pH 6.5 to 8
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.