Learn More
Alkali Scientific ASI™ Rabbit Serum, 500mL
SDP
Catalog No. 503028694
Encompass_Preferred
Rabbit serum is collected in France from animals typically six months old or younger. It is processed, and manufactured in the United States. Virus-Screened Mycoplasma-Tested Triple 0.2 Micron Filtered This product ships on ice.
Catalog No. 50-302-8694 Supplier Alkali Scientific Supplier No. FB31
Rabbit serum is collected in France from animals typically six months old or younger. It is processed, and manufactured in the United States. Virus-Screened Mycoplasma-Tested Triple 0.2 Micron Filtered This product ships on ice.