Learn More
Biomatik Corporation Rabbit Serum Amyloid A (SAA) ELISA Kit
Supplier:  Biomatik Corporation EKU10222
Encompass_Preferred
Rabbit Serum Amyloid A (SAA) ELISA Kit for detecting SAA in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.069ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P53614. Gene ID: 100009168. Target Synonyms: SA-A; SAA1; PIG4; TP53I4; Amyloid protein A; Amyloid fibril protein AA; Serum amyloid A-1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6229