Learn More
American Research Products Inc Rabbit SeX hormone binding glo
Catalog No. 5000328064
Encompass
Rabbit SeX hormone-binding globulin (SHBG) ELISA kit from CUSABIO Detection Range 25 pmol/mL-400 pmol/mL Sensitivity 25 pmol/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28064 Supplier American Research Products Inc Supplier No. CSBEL021252RB
Rabbit SeX hormone-binding globulin (SHBG) ELISA kit from CUSABIO Detection Range 25 pmol/mL-400 pmol/mL Sensitivity 25 pmol/mL Sample serum plasma tissue homogenates Method Competitive