Learn More
American Research Products Inc Rabbit Soluble protein 100B S
Catalog No. 5000329632
Encompass
Rabbit Soluble protein-100B S-100B ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29632 Supplier American Research Products Inc Supplier No. CSBE06895RB
Rabbit Soluble protein-100B S-100B ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich