Learn More
American Research Products Inc Rabbit stromal cell deriVed fa
Catalog No. 5000329730
Encompass
Rabbit stromal cell deriVed factor 1 (SDF-1/CXCL12) ELISA kit from CUSABIO Detection Range 0 625 ng/ml-10 ng/ml Sensitivity 0 125 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29730 Supplier American Research Products Inc Supplier No. CSBE12656RB
Rabbit stromal cell deriVed factor 1 (SDF-1/CXCL12) ELISA kit from CUSABIO Detection Range 0 625 ng/ml-10 ng/ml Sensitivity 0 125 ng/ml Sample serum plasma tissue homogenates Method Sandwich