Learn More
Biomatik Corporation Rabbit Superoxide Dismutase 1 (SOD1) ELISA Kit
Supplier:  Biomatik Corporation EKU08416
Encompass_Preferred
Rabbit Superoxide Dismutase 1 (SOD1) ELISA Kit for detecting SOD1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09212. Target Synonyms: ALS; ALS1; IPOA; Superoxide Dismutase 1, Soluble; Amyotrophic Lateral Sclerosis 1,Adult; Superoxide dismutase [Cu-Zn]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4856