Learn More
American Research Products Inc Rabbit Tartrate resistant acid
Catalog No. 5000328601
Encompass
Rabbit Tartrate-resistant acid phosphatase 5b (TRAP5b) ELISA Kit from CUSABIO Detection Range 78 U/mL-5000 U/mL Sensitivity 19 5 U/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28601 Supplier American Research Products Inc Supplier No. CSBE15043RB
Rabbit Tartrate-resistant acid phosphatase 5b (TRAP5b) ELISA Kit from CUSABIO Detection Range 78 U/mL-5000 U/mL Sensitivity 19 5 U/mL Sample serum plasma tissue homogenates Method Sandwich