Learn More
American Research Products Inc Rabbit terminal complement com
Catalog No. 5000329564
Encompass
Rabbit terminal complement compleX C5b-9 (TCC C5b-9) ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29564 Supplier American Research Products Inc Supplier No. CSBE15838RB
Rabbit terminal complement compleX C5b-9 (TCC C5b-9) ELISA Kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma Method Sandwich