Learn More
American Research Products Inc Rabbit Thyroid Stimulating Hor
Catalog No. 5000326672
Encompass
Rabbit Thyroid Stimulating Hormone TSH ELISA Kit from CUSABIO Detection Range 0 5 IU/mL-20 IU/mL Sensitivity 0 25 IU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26672 Supplier American Research Products Inc Supplier No. CSBE06916RB
Rabbit Thyroid Stimulating Hormone TSH ELISA Kit from CUSABIO Detection Range 0 5 IU/mL-20 IU/mL Sensitivity 0 25 IU/mL Sample serum plasma tissue homogenates Method Sandwich