Learn More
American Research Products Inc Rabbit tissue inhibitors of me
Catalog No. 5000329635
Encompass
Rabbit tissue inhibitors of metalloproteinase 1 TIMP-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive
Catalog No. 50-003-29635 Supplier American Research Products Inc Supplier No. CSBE06935RB
Rabbit tissue inhibitors of metalloproteinase 1 TIMP-1 ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Competitive