Learn More
Biomatik Corporation Rabbit Toll Like Receptor 9 (TLR9) ELISA Kit
Supplier:  Biomatik Corporation EKU09020
Encompass_Preferred
Rabbit Toll Like Receptor 9 (TLR9) ELISA Kit for detecting TLR9 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G4VV65. Target Synonyms: CD289. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6431