Learn More
American Research Products Inc Rabbit Transforming Growth fac
Catalog No. 5000326275
Encompass
Rabbit Transforming Growth factor b1 TGF-b1 ELISA Kit from CUSABIO Detection Range 3 125 ng/ml-50 ng/ml Sensitivity 0 625ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26275 Supplier American Research Products Inc Supplier No. CSBE07000RB
Rabbit Transforming Growth factor b1 TGF-b1 ELISA Kit from CUSABIO Detection Range 3 125 ng/ml-50 ng/ml Sensitivity 0 625ng/ml Sample serum plasma tissue homogenates Method Sandwich