Learn More
Rockland Immunochemicals Rabbit TrueBlot Set (with IP Agarose beads) - 88-1688-31
SDP
Supplier:  Rockland Immunochemicals 88168831
Encompass
Rabbit IgG TrueBlot is provided as 1000X solution. We recommend incubating the blots from minigels in sealed bags with minimal volume (2-3 mLs). If used conservatively at 2.5mls/blot will yield enough reagent for 20 blots. Note that there are three key procedural considerations: 1. Protein A or G should not be used for the IP. Use of protein A or G beads with the rabbit TrueBlot will result in contaminating bands. For IP, Anti-Rat IgG beads, or Anti-Rabbit IgG beads should be used for rat or rabbit immunoprecipitating antibodies, respectively. 2. Immunoprecipitate should be completely reduced. 3. BLOTTO/Milk should be used as the blocking protein for the immunoblot. Upon initial use of the IP beads, we recommend that the vial be inverted several times to get the beads into suspension. We recommend using a large bore pipet to pipet up the liquid for use. Specific conditions for each protein and antibody combination should be specifically optimized by the end user.
Catalog No. NC0412666