Learn More
American Research Products Inc Rabbit Tumor necrosis factor a
Catalog No. 5000330178
Encompass
Rabbit Tumor necrosis factor a TNFa ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-30178 Supplier American Research Products Inc Supplier No. CSBE06998RB
Rabbit Tumor necrosis factor a TNFa ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich