Learn More
XpressBio Rabbit Anti-Haemophilus influenzae B (HIB) ELISA Kit
SDP
Supplier:  XpressBio IM218
Encompass_Preferred
Quantitative ELISA assay developed to specifically measure the degree of immunity against a specific range of infectious targets. The microtiter wells coated with Haemophilus influenzae type b oligosaccharide-conjugated to human serum abumin (HbOHA) are exposed to test specimens, which may contain Rabbit IgG reactive antibodies.
Catalog No. 50-171-7665