Learn More
American Research Products Inc Rabbit Vascular Endothelial ce
Supplier:  American Research Products Inc CSBE08524RB
Encompass
Rabbit Vascular Endothelial cell Growth Factor VEGF ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26699