Learn More
Kingfisher Biotech Inc Rabbit VEGF-A 165 aa Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1025U025
Encompass
The Rabbit VEGF-A yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Rabbit VEGF-A applications are for cell culture, ELISA standard, and Western Blot Control. Rabbit VEGF-A yeast-derived recombinant protein can be purchased in multiple sizes. Rabbit VEGF-A Specifications: (Molecular Weight: 19.3 kDa) (Amino Acid Sequence: APMAEEGDNK PHEVVKFMEV YRRSYCQPIE TLVDIFQEYP DEIEYIFKPS CVPLVRCGGC CNDESLECVP TEEFNVTMQI MRIKPHQGQH IGEMSFLQHN KCECRPKKDR ARQENPCGPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRR (165)) (Gene ID: 100008899). For research use only. Made in the USA
Catalog No. 50-241-3930