Learn More
Immunoreagents Inc RABBITA-CHICKEN IGG AP 2 MG
Supplier:  Immunoreagents Inc RBXCK003D
Encompass_Preferred
Secondary Antibody; Rabbit anti-chicken IgG/Y (H&L), Affinity Pure, Host: Rabbit, Format: Liq, Label: None, Applications: WB, ELISA, IHC/ICC, IF, IP, FC
Catalog No. 50-270-851