Learn More
MyBioSource Monoclonal Antibody, Fungal Beta Glucan
SDP
Supplier:  MyBioSource MBS3134761 MG
Encompass_Preferred
The product is a monoclonal antibody that targets fungal beta-glucan. It is produced in mouse and can detect fungal beta-glucan in ELISA, Western Blot, Immunohistochemistry, and Flow Cytometry applications. The antibody reacts with beta-glucan from Candida, Aspergillus, Pneumocystis, Saccharomyces, and Cryptococcus. It does not cross-react with alpha-glucan. The clone number is BFDiv. The recommended dilution for ELISA is 0.5-2 μg/mL. This antibody can be a useful tool for detecting and studying fungal beta-glucan.
Catalog No. NC2743779