Learn More
Immunoreagents Inc RABBT A-HAMSTER IGG DYLIGHT633
Catalog No. 50272194
Encompass_Preferred
Secondary Antibody; Rabbit anti-hamster IgG (H&L), Affinity Pure, DyLight 633 Conjugate, Host: Rabbit, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS
Catalog No. 50-272-194 Supplier Immunoreagents Inc Supplier No. RBXHA003D633NHSX
Secondary Antibody; Rabbit anti-hamster IgG (H&L), Affinity Pure, DyLight 633 Conjugate, Host: Rabbit, Format: Lyo, Label: DyL633, Applications: WB, ELISA, IHC/ICC, IF, IP, FC, FACS