Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rabdosiin ((+)-Rabdosiin) | 263397-69-9 | 10 MG
SDP

Catalog No. 5000417717
Encompass_Preferred

Rabdosiin ((+)-Rabdosiin) is a Caffeic acid tetramer isolated from *Macrotomia euchroma*. It appears as an off-white to yellow solid.

  • Intended for research use only.
  • Molecular weight of 718.61.
  • Molecular formula C36H30O16.

Catalog No. 50-004-17717 Supplier Medchemexpress LLC Supplier No. HYN688010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabdosiin ((+)-Rabdosiin) is a Caffeic acid tetramer isolated from *Macrotomia euchroma*. It appears as an off-white to yellow solid.

  • Intended for research use only.
  • Molecular weight of 718.61.
  • Molecular formula C36H30O16.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.