Learn More
Medchemexpress LLC Rabeprazole-d4 sodium | 117976-90-6 | 98.6% | 385.45 | C18H16D4N3NaO3S | 10 MG
SDP
Catalog No. 5000298078
Encompass_Preferred
Rabeprazole-d4 sodium is the deuterium-labeled sodium salt of rabeprazole, supplied as a stable isotope internal standard for analytical assays. It is intended for quantitative mass spectrometry and other analytical methods that require an isotopically labeled reference.
  • Deuterium-labeled internal standard for LC-MS quantification.
  • High chemical purity for reliable analytical performance.
  • Suitable for pharmacokinetic and bioanalytical method development.
  • Supplied in a small-mass package for trace-level studies.
  • Stable under recommended storage conditions.
Catalog No. 50-002-98078 Supplier Medchemexpress LLC Supplier No. HYB0656AS10MG
Rabeprazole-d4 sodium is the deuterium-labeled sodium salt of rabeprazole, supplied as a stable isotope internal standard for analytical assays. It is intended for quantitative mass spectrometry and other analytical methods that require an isotopically labeled reference.
  • Deuterium-labeled internal standard for LC-MS quantification.
  • High chemical purity for reliable analytical performance.
  • Suitable for pharmacokinetic and bioanalytical method development.
  • Supplied in a small-mass package for trace-level studies.
  • Stable under recommended storage conditions.