Learn More
Medchemexpress LLC Rabeprazole sodium | 117976-90-6 | 99.9% | 1 ML
SDP
Catalog No. 5000408443
Encompass_Preferred
Rabeprazole sodium (LY307640 sodium) is provided as a 10 mM solution in 1 mL of DMSO. It is a second-generation proton pump inhibitor (PPI) that irreversibly inactivates gastric H+/K+-ATPase. This compound induces apoptosis and acts as an uridine nucleoside ribohydrolase (UNH) inhibitor with an IC50 of 0.3 μM. It is suitable for research on gastric ulcerations and gastroesophageal reflux and is intended for research use only.
  • Second-generation proton pump inhibitor (PPI)
  • Irreversibly inactivates gastric H+/K+-ATPase
  • Induces apoptosis
  • Acts as an uridine nucleoside ribohydrolase (UNH) inhibitor with an IC50 of 0.3 μM
  • Suitable for research on gastric ulcerations and gastroesophageal reflux
Catalog No. 50-004-08443 Supplier Medchemexpress LLC Supplier No. HYB0656A10MM/1ML
Rabeprazole sodium (LY307640 sodium) is provided as a 10 mM solution in 1 mL of DMSO. It is a second-generation proton pump inhibitor (PPI) that irreversibly inactivates gastric H+/K+-ATPase. This compound induces apoptosis and acts as an uridine nucleoside ribohydrolase (UNH) inhibitor with an IC50 of 0.3 μM. It is suitable for research on gastric ulcerations and gastroesophageal reflux and is intended for research use only.
  • Second-generation proton pump inhibitor (PPI)
  • Irreversibly inactivates gastric H+/K+-ATPase
  • Induces apoptosis
  • Acts as an uridine nucleoside ribohydrolase (UNH) inhibitor with an IC50 of 0.3 μM
  • Suitable for research on gastric ulcerations and gastroesophageal reflux