Learn More
Medchemexpress LLC HY-W003467 100mg Medchemexpress, Rabeprazole Sulfide CAS:117977-21-6 Purity:>98%
SDP
Catalog No. 502083215
Encompass_Preferred
Medchemexpress, HY-W003467 100mg Rabeprazole Sulfide CAS:117977-21-6 Rabeprazole Sulfide is an active metabolite of Rabeprazole. Rabeprazole is a proton pump inhibitor that suppresses gastric acid secretion through an interaction with (H+/K+)-ATPase in gastric parietal cells. Rabeprazole markedly inhibits the motility of H. pylori. Rabeprazole has the potential for various peptic diseases treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3215 Supplier Medchemexpress LLC Supplier No. HYW003467100MG
Medchemexpress, HY-W003467 100mg Rabeprazole Sulfide CAS:117977-21-6 Rabeprazole Sulfide is an active metabolite of Rabeprazole. Rabeprazole is a proton pump inhibitor that suppresses gastric acid secretion through an interaction with (H+/K+)-ATPase in gastric parietal cells. Rabeprazole markedly inhibits the motility of H. pylori. Rabeprazole has the potential for various peptic diseases treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.