Learn More
Cayman Chemical Rabeprazole Sulfide, Purity: ≥98%, CAS: 117977-21-6, 500 mg
SDP
Catalog No. 502911542
Encompass_Preferred
An active metabolite of rabeprazole; inhibits the motility of H. pylori (IC50 = 0.25 µg/ml); cytotoxic to HepG2 and PANC-1 cells (IC50s = 0.08 and 0.17 µM, respectively); a potential impurity in commercial preparations of rabeprazole; an acid and neutral hydrolytic degradation product of rabeprazole
Catalog No. 50-291-1542 Supplier Cayman Chemical Supplier No. 36276500 MG
An active metabolite of rabeprazole; inhibits the motility of H. pylori (IC50 = 0.25 µg/ml); cytotoxic to HepG2 and PANC-1 cells (IC50s = 0.08 and 0.17 µM, respectively); a potential impurity in commercial preparations of rabeprazole; an acid and neutral hydrolytic degradation product of rabeprazole