Learn More
Medchemexpress LLC Rabeprazole sulfone | 117976-47-3 | MFCD08063743 | 250mg
SDP
Catalog No. 5000267374
Encompass_Preferred
Rabeprazole sulfone is a metabolite of the proton pump inhibitor Rabeprazole and is an impurity reference for Rabeprazole[1]
Catalog No. 50-002-67374 Supplier Medchemexpress LLC Supplier No. HYZ2337250MG
Rabeprazole sulfone is a metabolite of the proton pump inhibitor Rabeprazole and is an impurity reference for Rabeprazole[1]