Learn More
Cayman Chemical Rabeprazole (sodium salt), Purity: ≥98%, CAS: 117976-90-6, 50 mg
SDP
Catalog No. 502968499
Encompass_Preferred
A proton pump inhibitor; inhibits the gastric H+/K+ ATPase (IC50 = 72 nM); activated more rapidly and over a greater pH range than other proton pump inhibitors; inhibits gastric acid secretion in pylorus-ligated rats and a rat model of gastric fistula at 30 mg/kg; inhibits the growth of several strains of H. pylori in vitro (MIC50s = 1.57-3.13 μg/mL)
Catalog No. 50-296-8499 Supplier Cayman Chemical Supplier No. 1493950 MG
A proton pump inhibitor; inhibits the gastric H+/K+ ATPase (IC50 = 72 nM); activated more rapidly and over a greater pH range than other proton pump inhibitors; inhibits gastric acid secretion in pylorus-ligated rats and a rat model of gastric fistula at 30 mg/kg; inhibits the growth of several strains of H. pylori in vitro (MIC50s = 1.57-3.13 μg/mL)