Learn More
Aviva Systems Biology Corp RABGEF1 Antibody - N-terminal region (ARP55073_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55073_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RABGEF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RABGEF1. Alternate names: FLJ32302, RABEX5, rabex-5ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61414