Learn More
Aviva Systems Biology Corp RABGGTA Antibody - N-terminal region (ARP49163_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP49163_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RABGGTA. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RABGGTA. Alternate names: PTAR3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53576