Learn More
Med Vet International Rabidbac Vet For Canine, Bacteria, Test Strips 10 Tests Per Kit
Catalog No. 502831170
Encompass_Preferred
RapidBac™ Vet is a lateral flow immunoassay for the detection of Gram-positive and Gram-negative bacteria in urine. Bufferless formation Expand your in-house testing capability Test, then treat, in the same office visit Results in 15 minutes No refrigeration required18 month expiration
Catalog No. 50-283-1170 Supplier Med Vet International Supplier No. RB731
RapidBac™ Vet is a lateral flow immunoassay for the detection of Gram-positive and Gram-negative bacteria in urine. Bufferless formation Expand your in-house testing capability Test, then treat, in the same office visit Results in 15 minutes No refrigeration required18 month expiration