Learn More
Medchemexpress LLC HY-P0285A 1mg Medchemexpress, Rabies Virus Glycoprotein (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0285A1MG
Encompass_Preferred
Medchemexpress, HY-P0285A 1mg Rabies Virus Glycoprotein (TFA) CAS: Rabies Virus Glycoprotein (TFA) is a 29-amino-acid cell penetrating peptide derived from a rabies virus glycoprotein that can cross the blood-brain barrier (BBB) and enter brain cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3033
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.