Learn More
ACROBiosystems Rabbit Trop-2 Protein, His Tag, 100 μg
Catalog No. NC3357279
Encompass_Preferred
This recombinant protein is a tumor-associated calcium signal transducer 2 (TROP-2) homolog expressed in human HEK293 cells. TROP-2 is a type-1 transmembrane glycoprotein related to EpCAM that acts as a signal transducer and may regulate carcinoma cell growth. This protein contains amino acids Gln 35 to Pro 279 (Accession # G1T6B6-1) and carries a polyhistidine tag at the C-terminus.
  • Expressed in human 293 cells (HEK293).
  • Purity of ≥95% by SDS-PAGE and ≥90% by SEC-MALS.
  • Calculated molecular weight of 29.5 kDa, migrating at 40-50 kDa under reducing conditions due to glycosylation.
  • Endotoxin levels <1.0 EU per μg.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 with trehalose as a protectant.
  • Shipped at ambient temperature; long-term storage recommended at -20°C or lower.
Catalog No. NC3357279 Supplier ACROBiosystems Supplier No. TR2R52H9100UG
This recombinant protein is a tumor-associated calcium signal transducer 2 (TROP-2) homolog expressed in human HEK293 cells. TROP-2 is a type-1 transmembrane glycoprotein related to EpCAM that acts as a signal transducer and may regulate carcinoma cell growth. This protein contains amino acids Gln 35 to Pro 279 (Accession # G1T6B6-1) and carries a polyhistidine tag at the C-terminus.
  • Expressed in human 293 cells (HEK293).
  • Purity of ≥95% by SDS-PAGE and ≥90% by SEC-MALS.
  • Calculated molecular weight of 29.5 kDa, migrating at 40-50 kDa under reducing conditions due to glycosylation.
  • Endotoxin levels <1.0 EU per μg.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 with trehalose as a protectant.
  • Shipped at ambient temperature; long-term storage recommended at -20°C or lower.