Learn More
Medchemexpress LLC Rabusertib (LY2603618) | 911222-45-2 | 99.98% | 436.30 | 1 ML
SDP
Catalog No. 5000361657
Encompass_Preferred
Rabusertib (LY2603618) is a potent and selective Chk1 inhibitor with an IC50 of 7 nM. It effectively reduces Chk1 autophosphorylation and abrogates the G2/M DNA damage checkpoint in cells treated with DNA damaging agents. Inhibition of Chk1 by Rabusertib leads to impaired DNA synthesis, elevated H2A.X phosphorylation, and premature entry into mitosis.
  • Potent and selective inhibitor of Chk1 (IC50 of 7 nM)
  • Reduces Chk1 autophosphorylation (EC50 of 430 nM)
  • Abrogates G2/M DNA damage checkpoint
  • Impairs DNA synthesis
  • Elevates H2A.X phosphorylation
  • Induces premature entry into mitosis
Catalog No. 50-003-61657 Supplier Medchemexpress LLC Supplier No. HY1472010MM/1ML
Rabusertib (LY2603618) is a potent and selective Chk1 inhibitor with an IC50 of 7 nM. It effectively reduces Chk1 autophosphorylation and abrogates the G2/M DNA damage checkpoint in cells treated with DNA damaging agents. Inhibition of Chk1 by Rabusertib leads to impaired DNA synthesis, elevated H2A.X phosphorylation, and premature entry into mitosis.
  • Potent and selective inhibitor of Chk1 (IC50 of 7 nM)
  • Reduces Chk1 autophosphorylation (EC50 of 430 nM)
  • Abrogates G2/M DNA damage checkpoint
  • Impairs DNA synthesis
  • Elevates H2A.X phosphorylation
  • Induces premature entry into mitosis