Learn More
Sino Biological RABV RABV-NP Protein 100ug (Sino Biological)
Supplier:  Sino Biological 40913V07E100
Encompass_Preferred
A DNA sequence encoding the Rabies virus (strain CVS-11) Nucleoprotein/NP (Q8JXF6) (Met1-450Ser) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-3505