Learn More
Medchemexpress LLC (Rac)-1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol sodium | 124011-52-5 | 100.0% | 100 MG
SDP
Catalog No. 5000438107
Encompass_Preferred
(Rac)-1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol (sodium) is an anionic phospholipid that can be used for drug delivery and liposome synthesis. It can induce Tregs specific for liposomes' cargo and is taken up by antigen-presenting cells mediated via complement component 1q (C1q).
  • Anionic phospholipid
  • Used for drug delivery and liposome synthesis
  • Liposomes containing this compound can induce antigen-specific Tregs
  • Taken up by antigen-presenting cells mediated via complement component 1q (C1q)
  • Can reduce plaque formation and increase plaque stability in atherosclerotic mice
Catalog No. 50-004-38107 Supplier Medchemexpress LLC Supplier No. HY112307A100MG
(Rac)-1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol (sodium) is an anionic phospholipid that can be used for drug delivery and liposome synthesis. It can induce Tregs specific for liposomes' cargo and is taken up by antigen-presenting cells mediated via complement component 1q (C1q).
  • Anionic phospholipid
  • Used for drug delivery and liposome synthesis
  • Liposomes containing this compound can induce antigen-specific Tregs
  • Taken up by antigen-presenting cells mediated via complement component 1q (C1q)
  • Can reduce plaque formation and increase plaque stability in atherosclerotic mice