Learn More
Medchemexpress LLC (Rac)-1,2-Distearoyl-sn-glycero-3-phosphatidylglycerol sodium | 124011-52-5 | 99.97% | 50 MG
SDP
Catalog No. 5000438386
Encompass_Preferred
(Rac)-1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol (sodium) is an anionic phospholipid. It can be studied for drug delivery and the synthesis of liposomes. Liposomes containing this compound can induce Tregs that are specific for their cargo. This compound is taken up by antigen-presenting cells through complement component 1q (C1q).
  • Anionic phospholipid.
  • Can be used for drug delivery and liposome synthesis.
  • Liposomes containing this compound can induce antigen-specific Tregs.
  • Taken up by antigen-presenting cells via complement component 1q (C1q).
Catalog No. 50-004-38386 Supplier Medchemexpress LLC Supplier No. HY112307A50MG
(Rac)-1,2-Distearoyl-sn-Glycero-3-Phosphatidylglycerol (sodium) is an anionic phospholipid. It can be studied for drug delivery and the synthesis of liposomes. Liposomes containing this compound can induce Tregs that are specific for their cargo. This compound is taken up by antigen-presenting cells through complement component 1q (C1q).
  • Anionic phospholipid.
  • Can be used for drug delivery and liposome synthesis.
  • Liposomes containing this compound can induce antigen-specific Tregs.
  • Taken up by antigen-presenting cells via complement component 1q (C1q).