Learn More
Medchemexpress LLC (Rac)-16:0 PI(5)P-d5 (diammonium) | 99.0% | 930.10 | 100 UG
SDP
Catalog No. 5000363045
Encompass_Preferred
This product is (Rac)-16:0 PI(5)P-d5 diammonium, a deuterium-labeled phosphatidylinositol 5-phosphate. It is an isotope-labeled compound used as a tracer and internal standard for quantitative analysis in research applications. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS techniques
  • Intended for research use only
Catalog No. 50-003-63045 Supplier Medchemexpress LLC Supplier No. HY146836S2100UG
This product is (Rac)-16:0 PI(5)P-d5 diammonium, a deuterium-labeled phosphatidylinositol 5-phosphate. It is an isotope-labeled compound used as a tracer and internal standard for quantitative analysis in research applications. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Functions as a tracer
  • Serves as an internal standard for quantitative analysis
  • Suitable for NMR, GC-MS, or LC-MS techniques
  • Intended for research use only