Learn More
Medchemexpress LLC Rac-3-hydroxyphenylglycine | 31932-87-3 | 99.6% | 167.16 g/mol | C8H9NO3 | 50 MG
SDP
Catalog No. 5000211390
Encompass_Preferred
(Rac)-3-Hydroxyphenylglycine is a racemic mixture of (S)- and (R)-3-hydroxyphenylglycine used as a research compound in metabotropic glutamate receptor (mGluR) studies. The (S)-enantiomer is a potent, selective mGluR1 agonist; the material is supplied as a high-purity solid intended for pharmacology, receptor assays, and analytical reference work.
  • High purity (99.58%) suitable for research applications.
  • Racemic mixture containing both S and R enantiomers.
  • Used as a reference compound in mGluR pharmacology and receptor assays.
  • Stable solid form with defined storage conditions for reliable results.
  • Available in small, accurately weighed quantities for assay work.
Catalog No. 50-002-11390 Supplier Medchemexpress LLC Supplier No. HY10084150MG
(Rac)-3-Hydroxyphenylglycine is a racemic mixture of (S)- and (R)-3-hydroxyphenylglycine used as a research compound in metabotropic glutamate receptor (mGluR) studies. The (S)-enantiomer is a potent, selective mGluR1 agonist; the material is supplied as a high-purity solid intended for pharmacology, receptor assays, and analytical reference work.
  • High purity (99.58%) suitable for research applications.
  • Racemic mixture containing both S and R enantiomers.
  • Used as a reference compound in mGluR pharmacology and receptor assays.
  • Stable solid form with defined storage conditions for reliable results.
  • Available in small, accurately weighed quantities for assay work.