Learn More
Medchemexpress LLC (rac)-3-o-methyl dopa-d3 | 1219173-95-1 | MFCD11983476 | 98.7% | 214.23 | C10H10D3NO4 | 5 MG
SDP
Catalog No. 5000262649
Encompass_Preferred
(rac)-3-O-Methyl DOPA-d3 is a deuterium-labeled, racemic form of 3-O-methyldopa supplied as a solid isotopically labeled standard for use as a tracer or internal standard in quantitative analytical techniques such as NMR, GC-MS, and LC-MS. It is an off-white to light yellow solid with high reported purity suitable for analytical workflows.
  • Isotopically labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity (98.7%) for reliable analytical performance.
  • Supplied as a stable solid, off-white to light yellow.
  • Recommended storage under nitrogen, away from moisture.
  • In solvent: stable at -80°C for 6 months and -20°C for 1 month.
  • Available in small research pack sizes suitable for tracer studies.
Catalog No. 50-002-62649 Supplier Medchemexpress LLC Supplier No. HY132404S5MG
(rac)-3-O-Methyl DOPA-d3 is a deuterium-labeled, racemic form of 3-O-methyldopa supplied as a solid isotopically labeled standard for use as a tracer or internal standard in quantitative analytical techniques such as NMR, GC-MS, and LC-MS. It is an off-white to light yellow solid with high reported purity suitable for analytical workflows.
  • Isotopically labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity (98.7%) for reliable analytical performance.
  • Supplied as a stable solid, off-white to light yellow.
  • Recommended storage under nitrogen, away from moisture.
  • In solvent: stable at -80°C for 6 months and -20°C for 1 month.
  • Available in small research pack sizes suitable for tracer studies.