Learn More
Medchemexpress LLC (Rac)-5-Hydroxymethyl Tolterodine | 200801-70-3 | 99.05% | C22H31NO2 | 1 G
SDP
Catalog No. 5000434020
Encompass_Preferred
(Rac)-5-Hydroxymethyl Tolterodine, an active metabolite of Tolterodine, acts as a muscarinic acetylcholine receptor (mAChR) antagonist. It exhibits Ki values ranging from 2.3 nM to 2.9 nM for M1-M5 receptors and is used in overactive bladder research.
  • Competitively inhibits carbachol-induced contraction in guinea-pig isolated urinary bladder strips (KB of 0.84 nM; pA2 of 9.14).
  • Shows significant binding to muscarinic receptors in most tissues (in vivo), with prolonged bladder duration.
  • Targets mAChR1, mAChR3, mAChR4, mAChR5.
  • Soluble in DMSO (100 mg/mL) and various aqueous/oil-based solutions (≥ 2.5 mg/mL).
Catalog No. 50-004-34020 Supplier Medchemexpress LLC Supplier No. HY765701G
(Rac)-5-Hydroxymethyl Tolterodine, an active metabolite of Tolterodine, acts as a muscarinic acetylcholine receptor (mAChR) antagonist. It exhibits Ki values ranging from 2.3 nM to 2.9 nM for M1-M5 receptors and is used in overactive bladder research.
  • Competitively inhibits carbachol-induced contraction in guinea-pig isolated urinary bladder strips (KB of 0.84 nM; pA2 of 9.14).
  • Shows significant binding to muscarinic receptors in most tissues (in vivo), with prolonged bladder duration.
  • Targets mAChR1, mAChR3, mAChR4, mAChR5.
  • Soluble in DMSO (100 mg/mL) and various aqueous/oil-based solutions (≥ 2.5 mg/mL).