Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-5-hydroxymethyl tolterodine hydrochloride | 250214-40-5 | C22H32ClNO2 | 100 MG
SDP

Catalog No. 5000434057
Encompass_Preferred

(Rac)-5-Hydroxymethyl Tolterodine hydrochloride is an active metabolite of Tolterodine, functioning as a muscarinic acetylcholine receptor (mAChR) antagonist. This compound is suitable for research related to overactive bladder.

  • Functions as a muscarinic acetylcholine receptor (mAChR) antagonist
  • Suitable for overactive bladder research
  • Molecular weight: 377.95
  • Chemical formula: C22H32ClNO2
  • Appears as an off-white to light yellow solid
  • Inhibits carbachol-induced contraction of guinea-pig isolated urinary bladder strips
  • Shows binding activity to muscarinic receptors

Catalog No. 50-004-34057 Supplier Medchemexpress LLC Supplier No. HY76570A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-5-Hydroxymethyl Tolterodine hydrochloride is an active metabolite of Tolterodine, functioning as a muscarinic acetylcholine receptor (mAChR) antagonist. This compound is suitable for research related to overactive bladder.

  • Functions as a muscarinic acetylcholine receptor (mAChR) antagonist
  • Suitable for overactive bladder research
  • Molecular weight: 377.95
  • Chemical formula: C22H32ClNO2
  • Appears as an off-white to light yellow solid
  • Inhibits carbachol-induced contraction of guinea-pig isolated urinary bladder strips
  • Shows binding activity to muscarinic receptors

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.