Learn More
Abcam rac-5,7-Dimethyltocol, internal standard for tocopherols and tocotrienols, 1ML
Catalog No. 502706069
Encompass_Preferred
MW 416.7 Da, Purity >95%. Internal standard for tocopherols and tocotrienols. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6069 Supplier Abcam Supplier No. AB1438791ML
MW 416.7 Da, Purity >95%. Internal standard for tocopherols and tocotrienols. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement