Learn More
Medchemexpress LLC (Rac)-ABT-202 dihydrochloride | 1258641-38-1 | 98.8% | 236.14 | C9H15Cl2N3 | 10MG
SDP
Catalog No. 5000237363
Encompass_Preferred
(Rac)-ABT-202 dihydrochloride is the racemic dihydrochloride salt of ABT-202, a small-molecule agonist of nicotinic acetylcholine receptors (nAChRs) with reported analgesic activity. Supplied as a light brown solid, it is intended for research applications involving nAChR modulation.
  • Agonist of nicotinic acetylcholine receptors with reported analgesic activity.
  • Racemic dihydrochloride salt for consistent bioactivity.
  • Solid, light brown to brown appearance for easy handling.
  • High purity (≈98.8%) suitable for research use.
  • Molecular weight 236.14 g/mol for dosing and calculations.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Store solid at 4°C sealed; store solutions at -80°C (up to 6 months) or -20°C (up to 1 month).
Catalog No. 50-002-37363 Supplier Medchemexpress LLC Supplier No. HY124540B10MG
(Rac)-ABT-202 dihydrochloride is the racemic dihydrochloride salt of ABT-202, a small-molecule agonist of nicotinic acetylcholine receptors (nAChRs) with reported analgesic activity. Supplied as a light brown solid, it is intended for research applications involving nAChR modulation.
  • Agonist of nicotinic acetylcholine receptors with reported analgesic activity.
  • Racemic dihydrochloride salt for consistent bioactivity.
  • Solid, light brown to brown appearance for easy handling.
  • High purity (≈98.8%) suitable for research use.
  • Molecular weight 236.14 g/mol for dosing and calculations.
  • Available as solid and as a 10 mM solution in DMSO for convenience.
  • Store solid at 4°C sealed; store solutions at -80°C (up to 6 months) or -20°C (up to 1 month).