Learn More
Medchemexpress LLC Rac-ABT-202 dihydrochloride | 1258641-38-1 | 98.8% | 236.14 | C9H15Cl2N3 | 1MG
SDP
Catalog No. 5000236735
Encompass_Preferred
(Rac)-ABT-202 dihydrochloride is the racemic dihydrochloride salt of ABT-202, a small-molecule agonist of nicotinic acetylcholine receptors used in neuroscience and analgesia research. Supplied as a light brown to brown solid for preclinical and in vitro studies, the compound has high solubility in DMSO and documented formulation options for aqueous dosing; storage recommendations are provided for both solid material and prepared solutions.
  • Racemic dihydrochloride salt of ABT-202.
  • Agonist of nicotinic acetylcholine receptors (nAChRs).
  • Light brown to brown solid suitable for research use.
  • Soluble in DMSO (33.33 mg/mL) and formulatable in aqueous vehicles.
  • Store solid at 4°C sealed, protect from moisture; solutions: -80°C (6 months), -20°C (1 month).
  • Available in small research sizes, for example 1 mg, 5 mg, and 10 mg.
Catalog No. 50-002-36735 Supplier Medchemexpress LLC Supplier No. HY124540B1MG
(Rac)-ABT-202 dihydrochloride is the racemic dihydrochloride salt of ABT-202, a small-molecule agonist of nicotinic acetylcholine receptors used in neuroscience and analgesia research. Supplied as a light brown to brown solid for preclinical and in vitro studies, the compound has high solubility in DMSO and documented formulation options for aqueous dosing; storage recommendations are provided for both solid material and prepared solutions.
  • Racemic dihydrochloride salt of ABT-202.
  • Agonist of nicotinic acetylcholine receptors (nAChRs).
  • Light brown to brown solid suitable for research use.
  • Soluble in DMSO (33.33 mg/mL) and formulatable in aqueous vehicles.
  • Store solid at 4°C sealed, protect from moisture; solutions: -80°C (6 months), -20°C (1 month).
  • Available in small research sizes, for example 1 mg, 5 mg, and 10 mg.