Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (Rac)-ABT-202 dihydrochloride | 1258641-38-1 | 98.8% | 236.14 g/mol | C9H15Cl2N3 | 10MM 1ML
SDP

Catalog No. 5000236049
Encompass_Preferred

(Rac)-ABT-202 dihydrochloride is the racemic dihydrochloride salt of ABT-202, a nicotinic acetylcholine receptor (nAChR) agonist with reported analgesic activity. It is supplied as a solid and as a 10 mM solution in DMSO for research use; typical purity is approximately 98.8%.

  • Racemic dihydrochloride form suitable for pharmacology studies.
  • Available as 10 mM solution in DMSO and multiple solid pack sizes.
  • High purity (≈98.8%), suitable for biochemical assays.
  • Molecular weight 236.14 g/mol and formula C9H15Cl2N3.
  • Store at 4°C sealed; in solvent store -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-36049 Supplier Medchemexpress LLC Supplier No. HY124540B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Rac)-ABT-202 dihydrochloride is the racemic dihydrochloride salt of ABT-202, a nicotinic acetylcholine receptor (nAChR) agonist with reported analgesic activity. It is supplied as a solid and as a 10 mM solution in DMSO for research use; typical purity is approximately 98.8%.

  • Racemic dihydrochloride form suitable for pharmacology studies.
  • Available as 10 mM solution in DMSO and multiple solid pack sizes.
  • High purity (≈98.8%), suitable for biochemical assays.
  • Molecular weight 236.14 g/mol and formula C9H15Cl2N3.
  • Store at 4°C sealed; in solvent store -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.