Learn More
TARGETMOL CHEMICALS INC RAC-ACOLBIFENE 5MG
SDP
Catalog No. 502577418
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Rac)-Acolbifene (EM-343) is the racemic form of Acolbifene. Acolbifene is a estrogen receptor antagonist. (Rac)-Acolbifene exhibits anti-estrogenic and estrogenic activities. (Rac)-Acolbifene contains a piperidine ring and exhibits good pharmacological profilerelative binding affinity (RBA)=380. purity: 98%
Catalog No. 50-257-7418 Supplier TARGETMOL CHEMICALS INC Supplier No. T126545MG
Also available in 1 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Rac)-Acolbifene (EM-343) is the racemic form of Acolbifene. Acolbifene is a estrogen receptor antagonist. (Rac)-Acolbifene exhibits anti-estrogenic and estrogenic activities. (Rac)-Acolbifene contains a piperidine ring and exhibits good pharmacological profilerelative binding affinity (RBA)=380. purity: 98%